missing translation for 'onlineSavingsMsg'
Learn More

7-(2-Aminoethylamino)-1-cyclopropyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic Acid (Ethylenediamine Compound), Mikromol™

Product Code. 17973946
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17973946 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17973946 Supplier Mikromol Supplier No. MM0018.04
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

7-(2-Aminoethylamino)-1-cyclopropyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic Acid (Ethylenediamine Compound)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 7-(2-Aminoethylamino)-1-cyclopropyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic Acid (Ethylenediamine Compound)
CAS 103222-12-4
Analyte or Component Names 7-(2-Aminoethylamino)-1-cyclopropyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic Acid (Ethylenediamine Compound)
Shipping Condition Room Temperature
Molecular Formula C15 H16 F N3 O3
Certifications/Compliance ISO 17025
Synonym Ciprofloxacin Hydrochloride Imp. C (EP), Ciprofloxacin Imp. C (EP), Enrofloxacin Imp. G (EP), Ciprofloxacin Ethylenediamine Analog, 7-[(2-Aminoethyl)amino]-1-cyclopropyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic Acid, 7-(2-Aminoethylamino)-1-cyclopropyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic Acid, Ethylenediamine Ciprofloxacin, Ciprofloxacin Hydrochloride Impurity C, Enrofloxacin Impurity G, Ciprofloxacin Impurity C
InChI Formula InChI=1S/C15H16FN3O3/c16-11-5-9-13(6-12(11)18-4-3-17)19(8-1-2-8)7-10(14(9)20)15(21)22/h5-8,18H,1-4,17H2,(H,21,22)
SMILES NCCNc1cc2N(C=C(C(=O)O)C(=O)c2cc1F)C3CC3
IUPAC Name 7-(2-aminoethylamino)-1-cyclopropyl-6-fluoro-4-oxoquinoline-3-carboxylic acid
Molecular Weight (g/mol) 305.3
Quantity 50 mg
Formula Weight 305.1176 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.