missing translation for 'onlineSavingsMsg'
Learn More

7,12-Dimethylbenz(a)Anthracene, SPEX CertiPrep™

Product Code. 15236146 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15236146 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15236146 Supplier SPEX Certiprep Supplier No. S1610

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
7,12-Dimethylbenz(a)anthracene57-97-61000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Packaging 2mL Amber Prescored Glass Ampule
CAS 57-97-6
CAS Min % 0.1
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
CAS Max % 0.1
Molecular Formula C20H16
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
InChI Key ARSRBNBHOADGJU-UHFFFAOYSA-N
SMILES CC1=C2C=CC3=CC=CC=C3C2=C(C4=CC=CC=C14)C
IUPAC Name 7,12-dimethylbenzo[a]anthracene
Solvent or Matrix MeCl:Benzene
Molecular Weight (g/mol) 256.348
PubChem CID 6001
Quantity 1 mL
ChEBI CHEBI:254496
Chemical Name or Material 7,12-Dimethylbenz(a)anthracene
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 7,12-Dimethylbenz(a)anthracene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Aspiration toxicity Category 1
  • Carcinogenicity Category 1A
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Germ cell mutagenicity Category 1B
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H310-Fatal in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H372-Causes damage to organs.
Precautionary Statement
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: dichloromethane, benzene

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.