missing translation for 'onlineSavingsMsg'
Learn More

7,12-Dimethylbenz[a]anthracene, 98%

Product Code. 10219840 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
5 g
Packaging:
Glass bottle
Unit Size:
100mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10219840 5 g Glass bottle 5g
10711525 100 mg Glass bottle 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10219840 Supplier Thermo Scientific Acros Supplier No. 408180050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 57-97-6 | C20H16 | 256.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-97-6
Molecular Formula C20H16
Molecular Weight (g/mol) 256.34
MDL Number MFCD00003600
InChI Key ARSRBNBHOADGJU-UHFFFAOYSA-N
Synonym dmba, 7,12-dimethylbenz a anthracene, 7,12-dimethylbenzanthracene, 9,10-dimethyl-1,2-benzanthracene, 7,12-dmba, 7,12-dimethylbenzo a anthracene, 7,12-dimethylbenzanthrancene, 6,7-dimethyl-1,2-benzanthracene, 1,4-dimethyl-2,3-benzphenanthrene
PubChem CID 6001
ChEBI CHEBI:254496
IUPAC Name 7,12-dimethylbenzo[a]anthracene
SMILES CC1=C2C=CC3=CC=CC=C3C2=C(C4=CC=CC=C14)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 122°C to 125°C
Color Green to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Beilstein 05, III, 2413
Merck Index 15, 3257
Formula Weight 256.34
Percent Purity ≥98.5%
Physical Form Crystalline Powder
Chemical Name or Material 7, 12-Dimethylbenz[a]anthracene, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 7,12-Dimethylbenz¢a!anthracene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 1A
Hazard Statement
  • H350-May cause cancer.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.