missing translation for 'onlineSavingsMsg'
Learn More

7,10-Dichloro-2-methoxybenzo[b][1,5]naphthyridine, TRC

Product Code. 30380213
Change view
Click to view available options
Quantity:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30380213 5 g
30382293 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30380213 Supplier Toronto Research Chemicals Supplier No. TRCD4354105G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6626-40-0
Molecular Formula C13H8Cl2N2O
Molecular Weight (g/mol) 279.12
Synonym 7,10-Dichloro-2-methoxy-pyrido[3,2-b]quinoline, 7,10-Dichloro-2-methoxypyrido[3,2-b]quinoline, NSC 59862
SMILES COc1ccc2nc3cc(Cl)ccc3c(Cl)c2n1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C13H8Cl2N2O/c1-18-11-5-4-9-13(17-11)12(15)8-3-2-7(14)6-10(8)16-9/h2-6H,1H3
Formula Weight 278.0014
Chemical Name or Material 7,10-Dichloro-2-methoxybenzo[b][1,5]naphthyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.