missing translation for 'onlineSavingsMsg'
Learn More

(6Z)-6-Octadecenoic Acid, TRC

Product Code. 30179516
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179516 100 mg 100mg
30179012 50 mg 50mg
30179300 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179516 Supplier Toronto Research Chemicals Supplier No. TRCO860070100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 593-39-5
Molecular Formula C18 H34 O2
Molecular Weight (g/mol) 282.46
Synonym 6-Octadecenoic acid, (6Z)- (9CI, ACI), (6Z)-6-Octadecenoic acid (ACI), 6-Octadecenoic acid, (Z)- (8CI), Petroselinic acid (6CI), (6Z)-Octadecenoic acid, (Z)-6-Octadecenoic acid, 6(Z)-Octadecenoic acid, cis-6-Octadecenoic acid, cis-Petroselinic acid, cis-δ6-Octadecenoic acid, Petroselic acid, δ6-cis-Octadecenoic acid
IUPAC Name (Z)-octadec-6-enoic acid
SMILES CCCCCCCCCCC\C=C/CCCCC(=O)O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C18H34O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h12-13H,2-11,14-17H2,1H3,(H,19,20)/b13-12-
Formula Weight 282.2559
Chemical Name or Material Petroselinic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.