missing translation for 'onlineSavingsMsg'
Learn More

(6R)-5,6,7,8-Tetrahydrobiopterin dihydrochloride

Product Code. 15743447 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 15743447

Brand: Thermo Scientific Acros 328871000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 69056-38-8 | C9H15N5O3·2HCl | 314.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69056-38-8
Molecular Formula C9H15N5O3·2HCl
Molecular Weight (g/mol) 314.17
InChI Key RKSUYBCOVNCALL-NTVURLEBSA-N
Synonym sapropterin dihydrochloride, biopten, r-2-amino-6-1r,2s-1,2-dihydroxypropyl-5,6,7,8-tetrahydropteridin-4 3h-one dihydrochloride, kuvan, sapropterin hydrochloride, unii-rg277lf5b3, biopten tn, sapropterin hcl, 6r-tetrahydrobiopterin hydrochloride
PubChem CID 636369
ChEBI CHEBI:32120
IUPAC Name (6R)-2-amino-6-[(1R,2S)-1,2-dihydroxypropyl]-5,6,7,8-tetrahydro-1H-pteridin-4-one;dihydrochloride
SMILES CC(C(C1CNC2=C(N1)C(=O)N=C(N2)N)O)O.Cl.Cl

Specifications

Melting Point 250.0°C to 255.0°C
Quantity 100 mg
Merck Index 15,8505
Solubility Information Solubility in water: soluble.
Formula Weight 314.17
Chemical Name or Material (6R)-5,6,7,8-Tetrahydrobiopterin dihydrochloride
compliance-icons
Product Identifier
  • (6R)-5,6,7,8-Tetrahydro-L-biopterin dihydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Tetrahydrobiopterindihydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.