missing translation for 'onlineSavingsMsg'
Learn More

6Beta-Hydroxycortisone, TRC

Product Code. 30393216
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393216 50 mg -
30390333 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30393216 Supplier Toronto Research Chemicals Supplier No. TRCH91845050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16355-28-5
Molecular Formula C21 H28 O6
Molecular Weight (g/mol) 376.44
Synonym Pregn-4-ene-3,11,20-trione, 6,17,21-trihydroxy-, (6β)-, Corticosterone, 11-dehydro-6β,17-dihydroxy- (6CI), Pregn-4-ene-3,11,20-trione, 6β,17,21-trihydroxy- (7CI,8CI), (6β)-6,17,21-Trihydroxypregn-4-ene-3,11,20-trione, 6β,17α,21-Trihydroxy-4-pregnene-3,11,20-trione, 6β-Hydroxycortisone, NSC 15457
IUPAC Name (6R,8S,9S,10R,13S,14S,17R)-6,17-dihydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-1,2,6,7,8,9,12,14,15,16-decahydrocyclopenta[a]phenanthrene-3,11-dione
SMILES C[C@]12CCC(=O)C=C1[C@H](O)C[C@H]3[C@@H]4CC[C@](O)(C(=O)CO)[C@@]4(C)CC(=O)[C@H]23

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C21H28O6/c1-19-5-3-11(23)7-14(19)15(24)8-12-13-4-6-21(27,17(26)10-22)20(13,2)9-16(25)18(12)19/h7,12-13,15,18,22,24,27H,3-6,8-10H2,1-2H3/t12-,13-,15+,18+,19-,20-,21-/m0/s1
Formula Weight 376.1886
Chemical Name or Material 6beta-Hydroxycortisone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.