missing translation for 'onlineSavingsMsg'
Learn More

6Beta-Hydroxy-17Beta-estradiol 17-Valerate, TRC

Product Code. 30398589
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30398589 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30398589 Supplier Toronto Research Chemicals Supplier No. TRCH94187525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6beta-Hydroxy-17beta-estradiol 17-Valerate
CAS 1313382-32-9
Molecular Formula C23H32O4
Quantity 25 mg
Synonym (6beta,17beta)-Estra-1,3,5(10)-triene-3,6,17-triol 17-Pentanoate, 6beta-Hydroxy-17beta-estradiol Valerate, (17beta)-Valeryloxyestra-1,3,5(10)-triene-3,6beta-diol Valerate
InChI Formula InChI=1S/C23H32O4/c1-3-4-5-22(26)27-21-9-8-19-17-13-20(25)18-12-14(24)6-7-15(18)16(17)10-11-23(19,21)2/h6-7,12,16-17,19-21,24-25H,3-5,8-11,13H2,1-2H3/t16-,17-,19+,20-,21+,23+/m1/s1
SMILES CCCCC(=O)O[C@H]1CC[C@H]2[C@@H]3C[C@@H](O)c4cc(O)ccc4[C@H]3CC[C@]12C
IUPAC Name [(6R,8R,9S,13S,14S,17S)-3,6-dihydroxy-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthren-17-yl] pentanoate
Molecular Weight (g/mol) 372.5
Formula Weight 372.2301
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.