missing translation for 'onlineSavingsMsg'
Learn More

6beta,7beta,15beta,16beta-Dimethylene-3beta,5beta-dihydroxyandrostan-17-one, TRC

Product Code. 30386415
Change view
Click to view available options
Quantity:
25 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30386415 1 g
30375231 25 mg
30368941 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30386415 Supplier Toronto Research Chemicals Supplier No. TRCD4728201G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82543-16-6
Molecular Formula C21 H30 O3
Molecular Weight (g/mol) 330.46
Synonym 17H-Dicyclopropa[6,7:15,16]cyclopenta[a]phenanthren-17-one, octadecahydro-3,5-dihydroxy-10,13-dimethyl-, (3S,5R,6R,7R,8R,9S,10R,13S,14S,15S,16S)-, 17H-Dicyclopropa[6,7:15,16]cyclopenta[a]phenanthren-17-one, octadecahydro-3,5-dihydroxy-10,13-dimethyl-, [3S-(3α,5α,6β,7β,8α,9β,10α,13α,14β,15β,16β)]-, 3β,5β-Dihydroxy-6β,7β, 15β,16β-dimethylene-5β-androst-17-one, 6β,7β, 15β,16β-Dimethylene-3β,5β-dihydroxyandostan-17-one
IUPAC Name (1R,2R,4R,5R,7S,10R,11S,14S,16S,18S,19S)-5,7-dihydroxy-10,14-dimethylhexacyclo[9.8.0.02,4.05,10.014,19.016,18]nonadecan-15-one
SMILES C[C@]12CC[C@H]3[C@@H]([C@H]4C[C@H]4[C@]5(O)C[C@@H](O)CC[C@]35C)[C@@H]1[C@@H]6C[C@@H]6C2=O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C21H30O3/c1-19-5-4-14-16(17(19)11-7-12(11)18(19)23)13-8-15(13)21(24)9-10(22)3-6-20(14,21)2/h10-17,22,24H,3-9H2,1-2H3/t10-,11+,12-,13-,14-,15+,16+,17-,19-,20+,21+/m0/s1
Formula Weight 330.2195
Chemical Name or Material 6 beta,7 beta, 15 beta, 16 beta-Dimethylene-3 beta, 5 beta-dihydroxyandrostan-17-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.