missing translation for 'onlineSavingsMsg'
Learn More

(6Beta,16Alpha,17Alpha)-16,17-Epoxy-6-methoxy-3,4-cyclo-16,17-dihydroabiraterone, TRC

Product Code. 30385301
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
1mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385301 5 mg 5mg
30385473 2.5 mg 2.5mg
30388375 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30385301 Supplier Toronto Research Chemicals Supplier No. TRCE5893455MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (6Beta,16Alpha,17Alpha)-16,17-Epoxy-6-methoxy-3,4-cyclo-16,17-dihydroabiraterone
Quantity 5 mg
Shipping Condition Room Temperature
Molecular Formula C25H33NO2
Formula Weight 379.25
Molecular Weight (g/mol) 379.54
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.