missing translation for 'onlineSavingsMsg'
Learn More

6alpha-(Methoxymethyl)androsta-1,4-dien-3,17-dione, TRC

Product Code. 30479403
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30479403 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30479403 Supplier Toronto Research Chemicals Supplier No. TRCM22604525MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6alpha-(Methoxymethyl)androsta-1,4-dien-3,17-dione
CAS 881896-95-3
Molecular Formula C21 H28 O3
Quantity 25 mg
Synonym Exemestane Impurity 16
InChI Formula InChI=1S/C21H28O3/c1-20-8-6-14(22)11-18(20)13(12-24-3)10-15-16-4-5-19(23)21(16,2)9-7-17(15)20/h6,8,11,13,15-17H,4-5,7,9-10,12H2,1-3H3/t13-,15+,16+,17+,20-,21+/m1/s1
SMILES COC[C@H]1C[C@H]2[C@@H]3CCC(=O)[C@@]3(C)CC[C@@H]2[C@@]4(C)C=CC(=O)C=C14
IUPAC Name (6S,8R,9S,10R,13S,14S)-6-(methoxymethyl)-10,13-dimethyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthrene-3,17-dione
Molecular Weight (g/mol) 328.445
Formula Weight 328.204
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.