missing translation for 'onlineSavingsMsg'
Learn More

6Alpha-Hydroxy Cortisol-d4, TRC

Product Code. 30389011
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30389011 1 mg
30398967 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30389011 Supplier Toronto Research Chemicals Supplier No. TRCH9223121MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6alpha-Hydroxy Cortisol-d4
Molecular Formula C21H26D4O6
Quantity 1 mg
Synonym 6α,17-Dihydroxy-corticosterone-d4, 6α,11β,17,21-Tetrahydroxy-pregn-4-ene-3,20-dione-d4, NSC 79099-d4
InChI Formula InChI=1S/C21H30O6/c1-19-5-3-11(23)7-14(19)15(24)8-12-13-4-6-21(27,17(26)10-22)20(13,2)9-16(25)18(12)19/h7,12-13,15-16,18,22,24-25,27H,3-6,8-10H2,1-2H3/t12-,13-,15+,16+,18-,19+,20+,21-/m1/s1/i9D2,16D,18D
SMILES OCC([C@@]1(O)[C@@]2(C)C([2H])([2H])[C@@](O)([2H])[C@@]3([2H])[C@]([C@@]2([H])CC1)([H])C[C@H](O)C4=CC(CC[C@@]43C)=O)=O
IUPAC Name (6S,8R,9S,10R,11S,13S,14R,17S)-6,11,17-trihydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-1,2,6,7,8,9,10,11,12,13,14,15,16,17-tetradecahydro-3H-cyclopenta[a]phenanthren-3-one-9,11,12,12-d4
Molecular Weight (g/mol) 382.48
Formula Weight 382.23
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.