missing translation for 'onlineSavingsMsg'
Learn More

6Alpha-Chloro-17-acetoxy Progesterone, TRC

Product Code. 30003512
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30003512 1 mg 1mg
30009858 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30003512 Supplier Toronto Research Chemicals Supplier No. TRCC3632801MG
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C23 H31 Cl O4

Specifications

CAS 2477-73-8
Quantity 1 mg
Synonym Pregn-4-ene-3,20-dione, 17-(acetyloxy)-6-chloro-, (6α)-, Pregn-4-ene-3,20-dione, 6α-chloro-17-hydroxy-, acetate (7CI,8CI), Progesterone, 6α-chloro-17-hydroxy-, acetate (6CI), (6α)-17-(Acetyloxy)-6-chloropregn-4-ene-3,20-dione, 17-Acetoxy-6α-chloropregn-4-ene-3,20-dione, 6α-Chloro-17-acetoxyprogesterone, 6α-Chloro-17-hydroxyprogesterone acetate, Hydromadinone acetate, NSC 33170, 6α-Chloro-3,20-dioxopregn-4-en-17-yl acetate, 6α-Chloro-17α-acetoxyprogesterone
InChI Formula InChI=1S/C23H31ClO4/c1-13(25)23(28-14(2)26)10-7-18-16-12-20(24)19-11-15(27)5-8-21(19,3)17(16)6-9-22(18,23)4/h11,16-18,20H,5-10,12H2,1-4H3/t16-,17+,18+,20+,21-,22+,23+/m1/s1
SMILES CC(=O)O[C@@]1(CC[C@H]2[C@@H]3C[C@H](Cl)C4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C)C(=O)C
IUPAC Name [(6S,8R,9S,10R,13S,14S,17R)-17-acetyl-6-chloro-10,13-dimethyl-3-oxo-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl] acetate
Molecular Weight (g/mol) 406.94
Formula Weight 406.1911
Chemical Name or Material 6alpha-Chloro-17alpha-acetoxyprogesterone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.