missing translation for 'onlineSavingsMsg'
Learn More

6Alpha,9Alpha-Difluoro-11Beta,21-dihydroxy-pregna-1,4,16-triene-3,20-dione, TRC

Product Code. 30387655
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30387655 10 mg
30387326 25 mg
30377560 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30387655 Supplier Toronto Research Chemicals Supplier No. TRCD45174510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1270039-29-6
Molecular Formula C21H24F2O4
Molecular Weight (g/mol) 378.41
Synonym (6alpha,11beta)-6,9-Difluoro-11,21-dihydroxy-pregna-1,4,16-triene-3,20-dione, (6alpha,11beta)-6,9-Difluoro-11,21-dihydroxypregna-1,4,16-triene-3,20-dione
IUPAC Name (6S,8S,9R,10S,11S,13S,14S)-6,9-difluoro-11-hydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-7,8,11,12,14,15-hexahydro-6H-cyclopenta[a]phenanthren-3-one
SMILES C[C@]12C[C@H](O)[C@@]3(F)[C@@H](C[C@H](F)C4=CC(=O)C=C[C@]34C)[C@@H]1CC=C2C(=O)CO

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C21H24F2O4/c1-19-9-18(27)21(23)14(12(19)3-4-13(19)17(26)10-24)8-16(22)15-7-11(25)5-6-20(15,21)2/h4-7,12,14,16,18,24,27H,3,8-10H2,1-2H3/t12-,14-,16-,18-,19-,20-,21-/m0/s1
Formula Weight 378.1643
Chemical Name or Material 6alpha,9alpha-Difluoro-11beta,21-dihydroxy-pregna-1,4,16-triene-3,20-dione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.