missing translation for 'onlineSavingsMsg'
Learn More

6Alpha,3'-p-Dihydroxy Paclitaxel (1mg/mL in Dimethyl Sulfoxide), TRC

Product Code. 30407086
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30407086 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30407086 Supplier Toronto Research Chemicals Supplier No. TRCKIT54211X1ML
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 157230-10-9
Molecular Formula C47 H51 N O16
Quantity 1 mL
Synonym (αR,βS)-β-(Benzoylamino)-α,4-dihydroxybenzenepropanoic Acid 6,12b-Bis(acetyloxy)-12-(benzoyloxy)-2a,3,4,4a,5,6,9,10,11,12,12a,12b-dodecahydro-3,4,11-trihydroxy-4a,8,13,13-tetramethyl-5-oxo-7,11-methano-1H-cyclodeca[3,4]benz[1,2-b]oxet-9-yl Ester
InChI Formula InChI=1S/C47H51NO16/c1-23-30(62-43(58)33(52)32(26-17-19-29(51)20-18-26)48-41(56)27-13-9-7-10-14-27)21-47(59)40(63-42(57)28-15-11-8-12-16-28)36-45(6,38(55)35(61-24(2)49)31(23)44(47,4)5)37(54)34(53)39-46(36,22-60-39)64-25(3)50/h7-20,30,32-37,39-40,51-54,59H,21-22H2,1-6H3,(H,48,56)/t30-,32-,33+,34-,35+,36-,37-,39+,40-,45-,46+,47+/m0/s1
SMILES CC(=O)O[C@H]1C(=O)[C@]2(C)[C@@H](O)[C@H](O)[C@H]3OC[C@@]3(OC(=O)C)[C@H]2[C@H](OC(=O)c4ccccc4)[C@]5(O)C[C@H](OC(=O)[C@H](O)[C@@H](NC(=O)c6ccccc6)c7ccc(O)cc7)C(=C1C5(C)C)C
Molecular Weight (g/mol) 885.9
Formula Weight 885.3208
Concentration or Composition (by Analyte or Components) 6alpha,3'-p-dihydroxy Paclitaxel 1 mg/mL
Chemical Name or Material 6alpha,3'-p-dihydroxy Paclitaxel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.