missing translation for 'onlineSavingsMsg'
Learn More

(6Alpha,11Beta,16Alpha,17Alpha)-6,9-Difluoro-17-[(2-furanylcarbonyl)oxy]-11-hydroxy-16-methyl-3-oxoandrosta-1,4-diene-17-carboxylic Acid, TRC

Product Code. 30380654
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380654 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30380654 Supplier Toronto Research Chemicals Supplier No. TRCD45025410MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (6alpha,11beta,16alpha,17alpha)-6,9-Difluoro-17-[(2-furanylcarbonyl)oxy]-11-hydroxy-16-methyl-3-oxoandrosta-1,4-diene-17-carboxylic Acid
CAS 397864-63-0
Purity Grade Notes HPLC
Molecular Formula C26H28F2O7
Quantity 10 mg
Synonym GW 694301X, (6S,8S,9R,10S,11S,13S,14S,16R,17R)-6,9-Difluoro-17-((furan-2-carbonyl)oxy)-11-hydroxy-10,13,16-trimethyl-3-oxo-6,7,8,9,10,11,12,13,14,15,16,17-dodecahydro-3H-cyclopenta[a]phenanthrene-17-carboxylic Acid
InChI Formula InChI=1S/C26H28F2O7/c1-13-9-15-16-11-18(27)17-10-14(29)6-7-23(17,2)25(16,28)20(30)12-24(15,3)26(13,22(32)33)35-21(31)19-5-4-8-34-19/h4-8,10,13,15-16,18,20,30H,9,11-12H2,1-3H3,(H,32,33)/t13-,15+,16+,18+,20+,23+,24+,25+,26+/m1/s1
SMILES C[C@H](C1)[C@](OC(C2=CC=CO2)=O)(C(O)=O)[C@]3(C)[C@]1([H])[C@]4([H])C[C@H](F)C5=CC(C=C[C@]5(C)[C@@]4(F)[C@@H](O)C3)=O
Molecular Weight (g/mol) 490.49
Formula Weight 490.1803
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.