missing translation for 'onlineSavingsMsg'
Learn More

666-15, MedChemExpress

Product Code. 16492696
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16492696

Brand: Medchem Express HY101120

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 666-15 is a potent and selective CREB inhibitor with an IC50 of 81 nM. 666-15 suppresses tumor growth in a breast cancer xenograft model.

666-15 is a potent and selective CREB inhibitor with an IC50 of 81 nM. 666-15 suppresses tumor growth in a breast cancer xenograft model.

Specifications

Chemical Name or Material 666-15
CAS 1433286-70-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C33H31Cl2N3O5
Quantity 25 mg
Solubility Information DMSO : 125 mg/mL (201.44 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(NC1=CC=C(Cl)C=C1O)C2=CC3=CC=CC=C3C=C2OCCNC(C4=C(OCCCN)C=C(C=CC=C5)C5=C4)=O.Cl
Molecular Weight (g/mol) 620.52
Formula Weight 620.52
Percent Purity 99.16%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.