Learn More
YSI™ 6051030 Pro Series ISE/Conductivity Cable
Description
Measures any ISE (pH, ORP, pH/ORP combo, ammonium, nitrate, chloride), conductivity, salinity, specific conductance, total dissolved solids and temperature
- Conductivity sensor is built into the cable
- Cables over 1-meter also come with the 603062 cable management kit
- The 6051030 cables only work with the YSI Pro Plus handheld
- Does not include ISE sensor 15-176-209 (order separately)
Specifications
Specifications
| For Use With (Application) | Conductivity, pH, Temp |
| Includes | Conductivity sensor |
| Length (English) | 393.7 in. |
| Length (Metric) | 10 m |