missing translation for 'onlineSavingsMsg'
Learn More

YSI™ 6051030 Pro Series ISE/Conductivity Cable

Product Code. 12973497
Change view
Click to view available options
Length (English):
157.4 in.
393.7 in.
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (English) unitSize
12973497 157.4 in. Each
12983497 393.7 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12973497 Supplier YSI™ Supplier No. 60510304

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cable for Pro Series conductivity/pH/temperature meter

Measures any ISE (pH, ORP, pH/ORP combo, ammonium, nitrate, chloride), conductivity, salinity, specific conductance, total dissolved solids and temperature

  • Conductivity sensor is built into the cable
  • Cables over 1-meter also come with the 603062 cable management kit
  • The 6051030 cables only work with the YSI Pro Plus handheld
  • Does not include ISE sensor 15-176-209 (order separately)

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Conductivity, pH, Temp
Includes Conductivity sensor
Length (English) 157.4 in.
Length (Metric) 4 m
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.