missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ 6 X DNA loading Dye
Click to view available options
Quantity:
4 mL
Unit Size:
4mL
Specifications
Specifications
| For Use With (Application) | PCR,Electrophoresis |
| Quantity | 4 mL |
| Storage Requirements | Store at room temparature. For long term storage, store at -20°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction