Learn More
Ohaus™ 6 x Centrifuge Tube Adapters, Frontier™ R-A12x2/18MI Rotors
Facilitate centrifugation of tubes smaller than the original, provided carrier space for Frontier™ R-A12x2/18MI Rotors
Brand:  Ohaus™ 30314900
Additional Details : Weight : 1.00000kg
Product Code. 15866732
  • £66.70 / Pack of 6
Estimated Shipment: 19-06-2024
to see stock.
Description
Description
  • Allows you to centrifuge tubes that are smaller than the space provided in the rotor carrier
  • For over 16000 x G
  • Suitable for use in Frontier™ R-A12x2/18MI Rotors
Specifications
Specifications
1 x 1.5 mL
Adapter, 1x1.5ml for over16000xg, 6/pk
2
R-A12x2/18MI Rotor
1