missing translation for 'onlineSavingsMsg'
Learn More

6-(Trifluoromethyl)pyridine-3-sulfonyl chloride, Thermo Scientific™

Product Code. 10494379
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10494379 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10494379 Supplier Thermo Scientific Maybridge Supplier No. MAY00261DA
 Restricted Item
Estimated Shipment: 15-07-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 959996-58-8
Packaging Amber Glass Bottle
Molecular Formula C6H3ClF3NO2S
Quantity 1 g
Synonym 6-trifluoromethyl pyridine-3-sulfonyl chloride, 6-trifluoromethyl-3-pyridinesulfonyl chloride, 3-pyridinesulfonylchloride,6-trifluoromethyl, chloro 6-trifluoromethyl 3-pyridyl sulfone, 3-pyridinesulfonyl chloride, 6-trifluoromethyl
InChI Key NSGLNIQAWVNZFB-UHFFFAOYSA-N
SMILES C1=CC(=NC=C1S(=O)(=O)Cl)C(F)(F)F
IUPAC Name 6-(trifluoromethyl)pyridine-3-sulfonyl chloride
Molecular Weight (g/mol) 245.6
PubChem CID 45594311
Formula Weight 245.6
Percent Purity 97%
Chemical Name or Material 6-(Trifluoromethyl)pyridine-3-sulfonyl chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.