missing translation for 'onlineSavingsMsg'
Learn More

6-(Trifluoromethyl)nicotinic acid, 97%

Product Code. 11374476
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11374476 1 g 1g
11384476 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11374476 Supplier Thermo Scientific Alfa Aesar Supplier No. L20054.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 231291-22-8 | C7H3F3NO2 | 190.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 231291-22-8
Molecular Formula C7H3F3NO2
Molecular Weight (g/mol) 190.10
MDL Number MFCD00792430
InChI Key JNYLMODTPLSLIF-UHFFFAOYSA-M
Synonym 6-trifluoromethyl nicotinic acid, 6-trifluoromethyl pyridine-3-carboxylic acid, 6-trifluoromethylnicotinic acid, 6-trifluoromethyl-nicotinic acid, 2-trifluoromethyl-5-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 6-trifluoromethyl, 2-trifluoromethyl pyridine-5-carboxylic acid, 6-trifluoromethyl-3-pyridinecarboxylic acid
PubChem CID 2777551
IUPAC Name 6-(trifluoromethyl)pyridine-3-carboxylic acid
SMILES [O-]C(=O)C1=CC=C(N=C1)C(F)(F)F

Specifications

Melting Point 191°C to 192°C
Quantity 1 g
Formula Weight 191.11
Percent Purity 97%
Chemical Name or Material 6-(Trifluoromethyl)nicotinic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.