missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ 6 TMR Tre

Click to view available options
Quantity:
2 mg
Unit Size:
2mg
Description
6 TMR Tre is a fluorescent trehalose conjugate for imaging bacterial cell envelope. Selectively labels mycobacterial trehalose monomycolate (TMM). Allows real-time visualization of peptidoglycan and monomycolate dynamics at the single-cell level. Suitable for use with confocal and super-resolution microscopy.
Specifications
Specifications
| Chemical Name or Material | 6-[4(3)-[3,6-bis(dimethylamino)xanthylium-9-yl]-3(4)-carboxybenzoyl]amino-6-deoxy-α,α-D-trehalose |
| Quantity | 2 mg |
| Target | Fluorescent Probes for Imaging Bacteria |
| Molecular Formula | C37H43N3O14 |
| Formula Weight | 753.75 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction