missing translation for 'onlineSavingsMsg'
Learn More

6-Thioguanine, MedChemExpress

Product Code. 16466316
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16466316 100 mg 100mg
16441706 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16466316 Supplier Medchem Express Supplier No. HY13765
 Restricted Item
Estimated Shipment: 28-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 6-Thioguanine (Thioguanine; 2-Amino-6-purinethiol) is an anti-leukemia and immunosuppressant agent, acts as an inhibitor of SARS and MERS coronavirus papain-like proteases (PLpros) and also potently inhibits USP2 activity, with IC50s of 25 μM and 40 μM for Plpros and recombinant human USP2, respectively.

6-Thioguanine (Thioguanine

Specifications

Chemical Name or Material 6-Thioguanine
Physical Form Solid
CAS 154-42-7
Color Light Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C5H5N5S
For Use With (Application) Cancer-programmed cell death
Synonym Thioguanine 2-Amino-6-purinethiol
Solubility Information DMSO : 10 mg/mL (59.81 mM; Need ultrasonic and warming)
SMILES SC1=NC(N)=NC2=C1N=CN2
Molecular Weight (g/mol) 167.19
Formula Weight 167.19
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.