missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MedChemExpress 6-Thio-2'-Deoxyguanosine
Specifications
Specifications
| Molecular Weight (g/mol) | 283.31 g/mol |
| Product Type | Reference compound |
| Content And Storage | 4°C, protect from light∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light) |
| Shipping Condition | Room temperature |
| Purity | 97.0% |
| Quantity | 10 mg |
| Solubility Information | DMSO : 50 mg/mL (176.49 mM∣Need ultrasonic) |
| For Use With (Application) | Cancer-programmed cell death |
| CAS | 789-61-7 |
| Synonym | 6-thio-dG∣ β-TGdR |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?