missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress 6-Thio-2'-Deoxyguanosine

Product Code. 16443616
Encompass_Preferred
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
This item is not returnable. View return policy

Product Code. 16443616

Brand: MedChemExpress HY18762

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 6-Thio-2'-Deoxyguanosine is a nucleoside analogue that can be incorporated into de novo-synthesized telomeres by telomerase.

Specifications

Molecular Weight (g/mol) 283.31 g/mol
Product Type Reference compound
Content And Storage 4°C, protect from light∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
Shipping Condition Room temperature
Purity 97.0%
Quantity 25 mg
Solubility Information DMSO : 50 mg/mL (176.49 mM∣Need ultrasonic)
For Use With (Application) Cancer-programmed cell death
CAS 789-61-7
Synonym 6-thio-dG∣ β-TGdR
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.