missing translation for 'onlineSavingsMsg'
Learn More

6-thien-2-ylnicotinic acid, Thermo Scientific™

Product Code. 10326384
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10326384 5 g 5g
10623245 250 mg 250mg
10418643 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10326384 Supplier Thermo Scientific Maybridge Supplier No. CC54801DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 179408-54-9
Molecular Formula C10H7NO2S
Molecular Weight (g/mol) 205.23
MDL Number MFCD06410384
InChI Key QYGFXNUFKLRCRB-UHFFFAOYSA-N
Synonym 6-thien-2-ylnicotinic acid, 6-thiophen-2-yl nicotinic acid, 6-2-thienyl nicotinic acid, 6-thiophen-2-yl pyridine-3-carboxylic acid, 6-2-thienyl pyridine-3-carboxylic acid, acmc-209efk, 6-thien-2ylnicotinic acid, 6-thiophen-2-yl-nicotinic acid, 6-thiophen-2-ylnicotinic acid, 2-thien-2-yl-5-pyridinecarboxylic acid
PubChem CID 15411144
SMILES OC(=O)C1=CN=C(C=C1)C1=CC=CS1

Specifications

Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 5 g
IUPAC Name 6-thiophen-2-ylpyridine-3-carboxylic acid
Formula Weight 205.24
Percent Purity 97%
Chemical Name or Material 6-Thien-2-ylnicotinic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.