missing translation for 'onlineSavingsMsg'
Learn More

6-Thien-2-ylnicotinaldehyde, 97%, Thermo Scientific™

Product Code. 10401894
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10401894 5 g 5g
10205854 250 mg 250mg
10697493 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10401894 Supplier Thermo Scientific Maybridge Supplier No. CC54804DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 834884-61-6
Molecular Formula C10H7NOS
Molecular Weight (g/mol) 189.232
MDL Number MFCD04115421
InChI Key TZMYWLDEGHJHDI-UHFFFAOYSA-N
Synonym 6-thien-2-ylnicotinaldehyde, 6-thiophen-2-yl nicotinaldehyde, 6-thiophen-2-yl pyridine-3-carbaldehyde, 6-thiophen-2-yl-pyridine-3-carbaldehyde, 6-thien-2-ylnicotinylaldehyde, 6-2-thienyl pyridine-3-carbaldehyde, 6-thien-2-ylpyridine-3-carboxaldehyde, 3-pyridinecarboxaldehyde,6-2-thienyl, 2-thien-2-ylpyridine-5-carboxaldehyde
PubChem CID 24229542
IUPAC Name 6-thiophen-2-ylpyridine-3-carbaldehyde
SMILES C1=CSC(=C1)C2=NC=C(C=C2)C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 189.24
Percent Purity 97%
Chemical Name or Material 6-Thien-2-ylnicotinaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.