missing translation for 'onlineSavingsMsg'
Learn More

6-(Tetrahydropyran-4-yloxy)nicotinic acid, 97%, Thermo Scientific™

Product Code. 11306815
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11306815 250 mg Amber Glass Bottle 250mg
10328063 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11306815 Supplier Thermo Scientific Maybridge Supplier No. CC58301CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 886851-55-4
Molecular Formula C11H13NO4
Molecular Weight (g/mol) 223.228
MDL Number MFCD09258833
InChI Key OLOSQIMMFCSJHD-UHFFFAOYSA-N
Synonym 6-oxan-4-yloxy pyridine-3-carboxylic acid, 6-tetrahydropyran-4-yloxy nicotinic acid, 6-tetrahydro-2h-pyran-4-yloxy nicotinic acid, 6-tetrahydro-2h-pyran-4-yl oxy nicotinic acid, 2-tetrahydro-2h-pyran-4-yloxy pyridine-5-carboxylic acid, 3-pyridinecarboxylicacid, 6-tetrahydro-2h-pyran-4-yl oxy, 6-oxan-4-yl oxy pyridine-3-carboxylic acid, 4-5-carboxypyridin-2-yl oxy tetrahydro-2h-pyran
PubChem CID 24229576
IUPAC Name 6-(oxan-4-yloxy)pyridine-3-carboxylic acid
SMILES C1COCCC1OC2=NC=C(C=C2)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 223.23
Percent Purity 97%
Chemical Name or Material 6-(Tetrahydropyran-4-yloxy)nicotinic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.