missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Pyrrolidin-1-ylpyridine-2-carboxylic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Synonym | 6-1-pyrrolidyl pyridine-2-carboxylic acid, 6-pyrrolidin-1-yl picolinic acid, 6-pyrrolidin-1-yl pyridine-2-carboxylic acid, 2-pyridinecarboxylicacid, 6-1-pyrrolidinyl, 6-pyrrolidinylpyridine-2-carboxylic acid, 6-1-pyrrolidyl pyridine-2-carboxylicacid, 2-pyridinecarboxylicacid,6-1-pyrrolidinyl, 6-1-pyrrolidinyl-2-pyridinecarboxylic acid |
IUPAC Name | 6-(pyrrolidin-1-yl)pyridine-2-carboxylic acid |
Formula Weight | 192.22 |
Percent Purity | 97% |
Chemical Name or Material | 6-Pyrrolidin-1-ylpyridine-2-carboxylic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'