missing translation for 'onlineSavingsMsg'
Learn More

6-Pyrrolidin-1-ylpyridine-2-carbaldehyde, 97%, Thermo Scientific™

Product Code. 10245314
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10245314 10 g 10g
10685155 250 mg 250mg
10119302 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10245314 Supplier Thermo Scientific Maybridge Supplier No. CC38004EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 230618-24-3
Molecular Formula C10H12N2O
Molecular Weight (g/mol) 176.22
MDL Number MFCD08435859
InChI Key SZOZYXCTOMKVTL-UHFFFAOYSA-N
PubChem CID 10821162
IUPAC Name 6-pyrrolidin-1-ylpyridine-2-carbaldehyde
SMILES O=CC1=NC(=CC=C1)N1CCCC1

Specifications

Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 10 g
Synonym 6-pyrrolidin-1-yl picolinaldehyde, 6-pyrrolidin-1-yl pyridine-2-carbaldehyde, 6-pyrrolidin-1-yl-picolinaldehyde, 6-pyrrolidinylpyridine-2-carbaldehyde, 6-1-pyrrolidinyl-2-pyridinecarboxaldehyde, 2-pyridinecarboxaldehyde,6-1-pyrrolidinyl, 2-pyridinecarboxaldehyde, 6-1-pyrrolidinyl
Formula Weight 176.22
Percent Purity 97%
Chemical Name or Material 6-Pyrrolidin-1-ylpyridine-2-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.