missing translation for 'onlineSavingsMsg'
Learn More

6-Piperidinopyridine-2-carboxylic acid, ≥97%, Thermo Scientific™

Product Code. 10119682
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10119682 1 g Amber Glass Bottle 1g
10662675 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10119682 Supplier Thermo Scientific Maybridge Supplier No. CC38101DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 868755-50-4
Molecular Formula C11H14N2O2
Molecular Weight (g/mol) 206.25
MDL Number MFCD08435860
InChI Key ZBNWTBPSTOBOGV-UHFFFAOYSA-N
Synonym 6-piperidinopyridine-2-carboxylic acid, 6-piperidin-1-yl picolinic acid, 6-piperidin-1-yl pyridine-2-carboxylic acid, 1-6-carboxypyridin-2-yl piperidine, 6-piperidylpyridine-2-carboxylic acid, 2-carboxy-6-piperidin-1-yl pyridine, 2-pyridinecarboxylicacid, 6-1-piperidinyl
PubChem CID 18524421
SMILES OC(=O)C1=NC(=CC=C1)N1CCCCC1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name 6-(piperidin-1-yl)pyridine-2-carboxylic acid
Formula Weight 206.24
Percent Purity 97%
Chemical Name or Material 6-Piperidinopyridine-2-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.