missing translation for 'onlineSavingsMsg'
Learn More

6-Piperidinopyridine-2-carbaldehyde, ≥97%, Thermo Scientific™

Catalog Number 10748065
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
859850-71-8
C11H14N2O
190.246
MFCD08543443
CFKVMXWXARUDQI-UHFFFAOYSA-N
6-piperidin-1-yl picolinaldehyde, 6-piperidinopyridine-2-carbaldehyde, 6-piperidin-1-yl pyridine-2-carbaldehyde, 2-formyl-6-piperidin-1-yl pyridine, 6-piperidylpyridine-2-carbaldehyde, 2-pyridinecarboxaldehyde,6-1-piperidinyl, 2-pyridinecarboxaldehyde, 6-1-piperidinyl
18525732
6-piperidin-1-ylpyridine-2-carbaldehyde
C1CCN(CC1)C2=CC=CC(=N2)C=O
This item is not returnable. View return policy
859850-71-8
C11H14N2O
190.246
MFCD08543443
CFKVMXWXARUDQI-UHFFFAOYSA-N
6-piperidin-1-yl picolinaldehyde, 6-piperidinopyridine-2-carbaldehyde, 6-piperidin-1-yl pyridine-2-carbaldehyde, 2-formyl-6-piperidin-1-yl pyridine, 6-piperidylpyridine-2-carbaldehyde, 2-pyridinecarboxaldehyde,6-1-piperidinyl, 2-pyridinecarboxaldehyde, 6-1-piperidinyl
18525732
6-piperidin-1-ylpyridine-2-carbaldehyde
C1CCN(CC1)C2=CC=CC(=N2)C=O
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 250 mg
Formula Weight 190.24
Percent Purity 97%
Chemical Name or Material 6-Piperidinopyridine-2-carbaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'