missing translation for 'onlineSavingsMsg'
Learn More

6-Piperidinonicotinic acid, 97%, Thermo Scientific™

Product Code. p-3075859
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10641435 250 mg 250mg
10112882 1 g 1g
10653085 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10641435 Supplier Thermo Scientific Maybridge Supplier No. MO07044CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 120800-50-2
Molecular Formula C11H14N2O2
Molecular Weight (g/mol) 206.25
MDL Number MFCD03791231
InChI Key QLPWWMSKVYYSEY-UHFFFAOYSA-N
Synonym 6-piperidin-1-yl nicotinic acid, 6-piperidinonicotinic acid, 6-piperidin-1-yl pyridine-3-carboxylic acid, 6-piperidin-1-ylnicotinic acid, 6-1-piperidinyl pyridine-3-carboxylic acid, acmc-20alxo, 6-piperidin-1-yl nicotinicacid, 6-piperidylpyridine-3-carboxylic acid, 3-pyridinecarboxylicacid, 6-1-piperidinyl
PubChem CID 2794761
SMILES OC(=O)C1=CN=C(C=C1)N1CCCCC1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
IUPAC Name 6-piperidin-1-ylpyridine-3-carboxylic acid
Formula Weight 206.24
Percent Purity 97%
Chemical Name or Material 6-Piperidinonicotinic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.