missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6 Phosphofructo 2 Kinase Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32163PCP
This item is not returnable.
View return policy
Description
6 Phosphofructo 2 Kinase Polyclonal antibody specifically detects 6 Phosphofructo 2 Kinase in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| 6 Phosphofructo 2 Kinase | |
| Polyclonal | |
| Western Blot, Immunoprecipitation | |
| 6PF-2-K/Fru-2,6-P2ase 3,6-bisphosphatase, 6-phosphofructo-2-kinase/ fructose-2,6-bisphosphatase, 6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 3,6-phosphofructo-2-kinase/fructose-2,6-bisphosphatase, FLJ37326, inducible 6-phosphofructo-2-kinase/fructose-2,6-bisphosphatase, IPFK2, Renal carcinoma antigen NY-REN-56 | |
| The immunogen this antibody was made to, maps to a region between residue 470 to 520 of human 6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 3 using the numbering given in entry NP_004557.1 | |
| 0.1 mL | |
| Endocrinology | |
| 5209 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| PerCP | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction