Learn More
Thermo Scientific Maybridge 6-Phenylnicotinonitrile, 10g, 97%
Brand: Thermo Scientific Maybridge CC21116EA
6-Phenylnicotinonitrile, 10g, 97%
Code : 31
Additional Details : CAS Number : 39065-54-8 Weight : 0.06000kg
Brand: Thermo Scientific Maybridge CC21116EA
6-Phenylnicotinonitrile, 10g, 97%
Code : 31
Additional Details : CAS Number : 39065-54-8 Weight : 0.06000kg