missing translation for 'onlineSavingsMsg'
Learn More

6-Phenoxy-3-pyridinamine, ≥95%, Thermo Scientific™

Product Code. 10314282
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10314282 10 g 10g
10292392 1 g 1g
10321825 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10314282 Supplier Thermo Scientific Maybridge Supplier No. CC19614EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 25194-67-6
Molecular Formula C11H10N2O
Molecular Weight (g/mol) 186.214
MDL Number MFCD01692449
InChI Key DETKIRMPBJPJRQ-UHFFFAOYSA-N
Synonym 6-phenoxy-3-pyridinamine, 3-amino-6-phenoxypyridine, 5-amino-2-phenoxypyridine, pyridine, 5-amino-2-phenoxy, 6-phenoxypyridine-3-amine, 3-pyridinamine,6-phenoxy, 3-pyridinamine, 6-phenoxy, 6-phenoxy-pyridin-3-ylamine, 6-phenoxy-3-pyridylamine, 2-phenoxy-5-aminopyridine
PubChem CID 212856
IUPAC Name 6-phenoxypyridin-3-amine
SMILES C1=CC=C(C=C1)OC2=NC=C(C=C2)N

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 10 g
Formula Weight 186.21
Percent Purity 95%
Chemical Name or Material 6-Phenoxy-3-pyridinamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.