missing translation for 'onlineSavingsMsg'
Learn More

6-(p-Toluidino)-2-naphthalenesulfonic Acid Sodium Salt, TRC

Product Code. 30456174
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30456174 250 mg 250mg
30451747 50 mg 50mg
30452736 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30456174 Supplier Toronto Research Chemicals Supplier No. TRCT536288250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53313-85-2
Molecular Formula C13 H12 N6 O2
Molecular Weight (g/mol) 284.273
Synonym Sodium 6-p-Toluidino-2-naphthalene Sulfonate, 6-[(4-Methylphenyl)amino]-2-naphthalenesulfonic Acid Monosodium Salt
IUPAC Name 2-N-(1H-indazol-5-yl)-6-N-methyl-3-nitropyridine-2,6-diamine
SMILES CNc1ccc(c(Nc2ccc3[nH]ncc3c2)n1)N(=O)=O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C13H12N6O2/c1-14-12-5-4-11(19(20)21)13(17-12)16-9-2-3-10-8(6-9)7-15-18-10/h2-7H,1H3,(H,15,18)(H2,14,16,17)
Formula Weight 284.102
Percent Purity >95%
Chemical Name or Material 6-(p-Toluidino)-2-naphthalenesulfonic Acid Sodium Salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.