missing translation for 'onlineSavingsMsg'
Learn More

6-Oxo-1,6-dihydropyridazine-3-carboxylic acid monohydrate, 97%, Thermo Scientific™

Product Code. 10304372
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10304372 10 g Amber Glass Bottle 10g
10661473 1 g Amber Glass Bottle 1g
10326394 25 g Amber Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10304372 Supplier Thermo Scientific Maybridge Supplier No. BTB07063EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 37972-69-3
Molecular Formula C5H3N2O3
Molecular Weight (g/mol) 139.09
MDL Number MFCD09064936
InChI Key GIFSROMQVPUQFK-UHFFFAOYSA-M
Synonym 6-hydroxypyridazine-3-carboxylic acid, 6-hydroxy-3-pyridazinecarboxylic acid, 6-oxo-1,6-dihydropyridazine-3-carboxylic acid, 6-oxo-1,6-dihydro-pyridazine-3-carboxylic acid, 3-hydroxypyridazine-6-carboxylic acid, 3-pyridazinecarboxylic acid, 1,6-dihydro-6-oxo, 6-hydroxy-pyridazine-3-carboxylic acid, 6-oxo-1,6-dihydro-3-pyridazinecarboxylic acid, 3-pyridazinecarboxylicacid, 1,6-dihydro-6-oxo, 6-oxohydropyridazine-3-carboxylic acid
PubChem CID 305970
IUPAC Name 6-oxo-1H-pyridazine-3-carboxylic acid
SMILES [O-]C(=O)C1=NNC(=O)C=C1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 158.12
Percent Purity 97%
Chemical Name or Material 6-Hydroxy-3-pyridazinecarboxylic acid monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.