missing translation for 'onlineSavingsMsg'
Learn More

6-Oxo-1,4,5,6-tetrahydropyridazine-3-carboxylic acid, 97%, Thermo Scientific™

Product Code. p-2703449
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
This item is not returnable. View return policy

Product Code. 10522622

Brand: Thermo Scientific Maybridge BTB07054DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 27372-38-9
Molecular Formula C5H6N2O3
Molecular Weight (g/mol) 142.114
MDL Number MFCD00052030
InChI Key VUADWGRLHPTYPI-UHFFFAOYSA-N
Synonym 6-oxo-1,4,5,6-tetrahydropyridazine-3-carboxylic acid, 1,4,5,6-tetrahydro-6-oxopyridazine-3-carboxylic acid, unii-ot09ztp07e, ot09ztp07e, 6-oxo-1,4,5,6-tetrahydro-pyridazine-3-carboxylic acid, 3-pyridazinecarboxylic acid, 1,4,5,6-tetrahydro-6-oxo, 1,4,5,6-tetrahydro-6-oxo-3-pyridazinecarboxylic acid, 6-oxo-1,4,5,6-tetrahydropyridazin-3-carboxylic acid, 6-oxo-1,4,5-trihydropyridazine-3-carboxylic acid, pubchem19274
PubChem CID 99621
IUPAC Name 6-oxo-4,5-dihydro-1H-pyridazine-3-carboxylic acid
SMILES C1CC(=O)NN=C1C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 1 g
Formula Weight 142.11
Percent Purity 97%
Chemical Name or Material 6-Oxo-1,4,5,6-tetrahydropyridazine-3-carboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.