missing translation for 'onlineSavingsMsg'
Learn More

6-OAU, MedChemExpress

Product Code. 16438047
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16438047

missing translation for 'mfr': Medchem Express HY12764

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 6-OAU (GTPL5846) (6-n-octylaminouracil) is an GPR84 (G protein-coupled receptor 84) agonist, with an EC50 value of 105 nM. 6-OAU works as a chemoattractant to both PMNs and macrophages, and amplifies the proinflammatory cytokine IL-8, shows proinflammatory function. 6-OAU also displays anti-bacterial function.

6-OAU (GTPL5846) (6-n-octylaminouracil) is an GPR84 (G protein-coupled receptor 84) agonist, with an EC50 value of 105 nM. 6-OAU works as a chemoattractant to both PMNs and macrophages, and amplifies the proinflammatory cytokine IL-8, shows proinflammatory function. 6-OAU also displays anti-bacterial function.

Specifications

Chemical Name or Material 6-OAU
Physical Form Solid
CAS 83797-69-7
Color Light Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C12H21N3O2
For Use With (Application) COVID-19-immunoregulation
Synonym GTPL5846
Solubility Information DMSO : 25 mg/mL (104.47 mM; Need ultrasonic)
SMILES O=C1NC(C=C(NCCCCCCCC)N1)=O
Molecular Weight (g/mol) 239.31
Formula Weight 239.31
Percent Purity 99.94%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.