missing translation for 'onlineSavingsMsg'
Learn More

6-O-α-Maltosyl-β-cyclodextrin, MedChemExpress

Product Code. p-200096385
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 16491729

Brand: Medchem Express HY18593

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 6-O-α-Maltosyl-β-cyclodextrin is a cellular cholesterol modifier which can form soluble inclusion complex with cholesterol.

6-O-α-Maltosyl-β-cyclodextrin is a cellular cholesterol modifier which can form soluble inclusion complex with cholesterol.

Specifications

Chemical Name or Material 6-O-α-Maltosyl-β-cyclodextrin
CAS 104723-60-6
Color White
Purity Grade Notes Research
Molecular Formula C54H90O45
Quantity 1 g
Synonym 6-O-alpha-D-Maltosyl-beta-cyclodextrin
Solubility Information DMSO : ≥ 100 mg/mL (68.53 mM) ∣H2O : ≥ 60.5 mg/mL (41.46 mM)
SMILES O[C@@H]1[C@@H](CO)O[C@H](O[C@H]2[C@H](O)[C@@H](O)[C@@H](OC[C@H]3O[C@H](O[C@@H]4[C@@H](CO)O[C@H](O[C@@H]5[C@@H](CO)O[C@H](O6)[C@H](O)[C@H]5O)[C@H](O)[C@H]4O)[C@H](O)[C@@H](O)[C@@H]3O[C@H]7O[C@H](CO)[C@@H](O[C@@H]8[C@H](O)[C@@H](O)[C@H](O[C@@H]9[C@H](O)[C@@H](O)[C@H](OC%10[C@H](O)[C@@H](O)[C@H]6[C@@H](CO)O%10)[C@@H](CO)O9)[C@@H](CO)O8)[C@H](O)[C@H]7O)O[C@@H]2CO)[C@H](O)[C@H]1O
Molecular Weight (g/mol) 1459.27
Formula Weight 1459.27
Percent Purity 97.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.