missing translation for 'onlineSavingsMsg'
Learn More

6-O-α-Maltosyl-β-cyclodextrin, MedChemExpress

Product Code. 16491729
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16491729 1 g 1g
16489997 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16491729 Supplier Medchem Express Supplier No. HY18593
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress 6-O-α-Maltosyl-β-cyclodextrin is a cellular cholesterol modifier which can form soluble inclusion complex with cholesterol.

6-O-α-Maltosyl-β-cyclodextrin is a cellular cholesterol modifier which can form soluble inclusion complex with cholesterol.

Specifications

Chemical Name or Material 6-O-α-Maltosyl-β-cyclodextrin
CAS 104723-60-6
Color White
Purity Grade Notes Research
Molecular Formula C54H90O45
Quantity 1 g
Synonym 6-O-alpha-D-Maltosyl-beta-cyclodextrin
Solubility Information DMSO : ≥ 100 mg/mL (68.53 mM) ∣H2O : ≥ 60.5 mg/mL (41.46 mM)
SMILES O[C@@H]1[C@@H](CO)O[C@H](O[C@H]2[C@H](O)[C@@H](O)[C@@H](OC[C@H]3O[C@H](O[C@@H]4[C@@H](CO)O[C@H](O[C@@H]5[C@@H](CO)O[C@H](O6)[C@H](O)[C@H]5O)[C@H](O)[C@H]4O)[C@H](O)[C@@H](O)[C@@H]3O[C@H]7O[C@H](CO)[C@@H](O[C@@H]8[C@H](O)[C@@H](O)[C@H](O[C@@H]9[C@H](O)[C@@H](O)[C@H](OC%10[C@H](O)[C@@H](O)[C@H]6[C@@H](CO)O%10)[C@@H](CO)O9)[C@@H](CO)O8)[C@H](O)[C@H]7O)O[C@@H]2CO)[C@H](O)[C@H]1O
Molecular Weight (g/mol) 1459.27
Formula Weight 1459.27
Percent Purity 97.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.