missing translation for 'onlineSavingsMsg'
Learn More

6-O-alpha-D-Maltosyl-beta-cyclodextrin, TRC

Product Code. 17939654
Change view
Click to view available options
Quantity:
1 g
25 g
100 g
Unit Size:
100g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17939654 25 g 25g
17909644 100 g 100g
17937164 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17939654 Supplier Toronto Research Chemicals Supplier No. TRCM15857025G
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104723-60-6
Molecular Formula C54 H90 O45
Molecular Weight (g/mol) 1459.26
Synonym Maltosyl-beta-cyclodextrin, alpha-Maltosyl-beta-cyclodextrin
SMILES OC[C@H]1O[C@H](O[C@H]2[C@H](O)[C@@H](O)[C@@H](OCC3O[C@@H]4O[C@H]5[C@H](O)[C@@H](O)[C@H](O[C@@H]5CO)O[C@H]6[C@H](O)[C@@H](O)[C@H](O[C@@H]6CO)O[C@H]7[C@H](O)[C@@H](O)[C@H](O[C@@H]7CO)O[C@H]8[C@H](O)[C@@H](O)[C@H](O[C@@H]8CO)O[C@H]9[C@H](O)[C@@H](O)[C@H](O[C@@H]9CO)O[C@H]%10[C@H](O)[C@@H](O)[C@H](O[C@@H]%10CO)O[C@H]3[C@H](O)[C@H]4O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Quantity 25 g
InChI Formula InChI=1S/C54H90O45/c55-1-10-19(63)20(64)29(73)47(83-10)92-38-11(2-56)84-46(30(74)21(38)65)82-9-18-45-28(72)37(81)54(91-18)98-44-17(8-62)89-52(35(79)26(44)70)96-42-15(6-60)87-50(33(77)24(42)68)94-40-13(4-58)85-48(31(75)22(40)66)93-39-12(3-57)86-49(32(76)23(39)67)95-41-14(5-59)88-51(34(78)25(41)69)97-43-16(7-61)90-53(99-45)36(80)27(43)71/h10-81H,1-9H2/t10-,11-,12-,13-,14-,15-,16-,17-,18?,19-,20+,21-,22-,23-,24-,25-,26-,27-,28-,29-,30-,31-,32-,33-,34-,35-,36-,37-,38-,39-,40-,41-,42-,43-,44-,45-,46+,47-,48-,49-,50-,51-,52-,53-,54-/m1/s1
Formula Weight 1458.4754
Chemical Name or Material 6-O-Alpha-D-Maltosyl-Beta-cyclodextrin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.