missing translation for 'onlineSavingsMsg'
Learn More

6-Nitroveratraldehyde, 96%

Product Code. 11427047
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11427047 5 g 5g
11394629 10 g 10g
11437047 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11427047 Supplier Thermo Scientific Alfa Aesar Supplier No. A14415.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 20357-25-9 | C9H9NO5 | 211.17 g/mol

6-Nitroveratraldehyde has been used in the preparation of no-carrier-added 6-18F-fluoro-L-dopa, fundamental tracer for cerebral positron emission tomography studies of dopaminergic system in humans and o-nitroaryl-bis(5-methylfur-2-yl)methanes, versatile synthons for the synthesis of nitrogen-containing heterocycles. DMNB is also an enzyme involved in the non-homologous end-joining (NHEJ) pathway of double-stranded DNA break (DSB) repair. It is also used for synthesizing analogs of alpha-asarone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Nitroveratraldehyde has been used in the preparation of no-carrier-added 6-18F-fluoro-L-dopa, fundamental tracer for cerebral positron emission tomography studies of dopaminergic system in humans and o-nitroaryl-bis(5-methylfur-2-yl)methanes, versatile synthons for the synthesis of nitrogen-containing heterocycles. DMNB is also an enzyme involved in the non-homologous end-joining (NHEJ) pathway of double-stranded DNA break (DSB) repair. It is also used for synthesizing analogs of alpha-asarone.

Solubility
Insoluble in water. Soluble to 25 mM in ethanol with gentle warming and to 100 mM in DMSO.

Notes
Air and light sensitive. Store away from air, light and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20357-25-9
Molecular Formula C9H9NO5
Molecular Weight (g/mol) 211.17
MDL Number MFCD00007134
InChI Key YWSPWKXREVSQCA-UHFFFAOYSA-N
Synonym 6-nitroveratraldehyde, dmnb, 3,4-dimethoxy-6-nitrobenzaldehyde, 4,5-dimethoxy-2-nitro-benzaldehyde, dna-pk inhibitor, benzaldehyde, 4,5-dimethoxy-2-nitro, 2-nitro-4,5-dimethoxybenzaldehyde, dna-dependent protein kinase inhibitor, 2-nitro-4,5-bis methyloxy benzaldehyde, pubchem8213
PubChem CID 88505
IUPAC Name 4,5-dimethoxy-2-nitrobenzaldehyde
SMILES COC1=CC(C=O)=C(C=C1OC)[N+]([O-])=O

Specifications

Melting Point 131°C to 135°C
Quantity 5 g
Beilstein 395599
Sensitivity Air and light sensitive
Solubility Information Insoluble in water. Soluble to 25 mM in ethanol with gentle warming and to 100 mM in DMSO.
Formula Weight 211.17
Percent Purity 96%
Chemical Name or Material 6-Nitroveratraldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.