missing translation for 'onlineSavingsMsg'
Learn More

6-Nitrophthalide, 97%, ACROS Organics™

Product Code. 10061751 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
1g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10061751 1g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10061751 Supplier Thermo Scientific Acros Supplier No. 331950010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 610-93-5
CAS Min % 96.0
CAS Max % 100.0
Melting Point 140°C to 145°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C8H5NO4
MDL Number MFCD00033529
Quantity 1 g
Beilstein 17, 313
Synonym 6-nitrophthalide, 6-nitroisobenzofuran-1 3h-one, 6-nitro-3h-isobenzofuran-1-one, 6-nitro-1,3-dihydroisobenzofuran-1-one, 6-nitro-1 3h-isobenzofuranone, 6-nitro-2-benzofuran-1 3h-one, 1 3h-isobenzofuranone, 6-nitro, 6-nitro-3-hydroisobenzofuran-1-one, 6-nitro-1,3-dihydro-2-benzofuran-1-one, 6-nitrophtalide
InChI Key RNWGZXAHUPFXLL-UHFFFAOYSA-N
SMILES [O-][N+](=O)C1=CC=C2COC(=O)C2=C1
IUPAC Name 6-nitro-1,3-dihydro-2-benzofuran-1-one
Molecular Weight (g/mol) 179.13
PubChem CID 223584
Formula Weight 179.13
Percent Purity ≥96.0%
Physical Form Crystalline Powder
Chemical Name or Material 6-Nitrophthalide, 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.