missing translation for 'onlineSavingsMsg'
Learn More

6-Nitroindoline, 98%, Thermo Scientific™

Product Code. 10768942 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10768942 5 g Glass bottle 5g
10255871 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10768942 Supplier Thermo Scientific Acros Supplier No. 128710050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 19727-83-4
Molecular Formula C8H8N2O2
Molecular Weight (g/mol) 164.16
MDL Number MFCD00005710
InChI Key LTNYDSMDSLOMSM-UHFFFAOYSA-N
Synonym 6-nitroindoline, indoline, 6-nitro, 1h-indole, 2,3-dihydro-6-nitro, 6-nitro-indoline, 2,3-dihydro-6-nitro-1h-indole, ccris 3302, zlchem 357, pubchem7443
PubChem CID 29757
IUPAC Name 6-nitro-2,3-dihydro-1H-indole
SMILES [O-][N+](=O)C1=CC=C2CCNC2=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 66°C to 70°C
Color Orange or Red
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: negligible. Other solubilities: soluble in acetone,ethyl acetate and methanol
Formula Weight 164.16
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 6-Nitroindoline, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.