missing translation for 'onlineSavingsMsg'
Learn More

6-Nitro-4-quinazolone, TRC

Product Code. 30483548
Change view
Click to view available options
Quantity:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30483548 100 mg
30478447 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30483548 Supplier Toronto Research Chemicals Supplier No. TRCN596450100MG
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6943-17-5
Molecular Formula C8H5N3O3
Molecular Weight (g/mol) 191.14
Synonym 6-Nitro-3H-quinazolin-4-one, 6-Nitro-4(1H)-quinazolinone, 6-Nitroquinazolin-4-ol, 6-Nitro-3,4-dihydroquinazolin-4-one, NSC 51669
IUPAC Name 6-nitro-1H-quinazolin-4-one
SMILES [O-][N+](=O)c1ccc2NC=NC(=O)c2c1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C8H5N3O3/c12-8-6-3-5(11(13)14)1-2-7(6)9-4-10-8/h1-4H,(H,9,10,12)
Formula Weight 191.0331
Chemical Name or Material 6-Nitro-4-quinazolone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.