missing translation for 'onlineSavingsMsg'
Learn More

6-Nitro-2H-chromen-2-one, 97%, Maybridge

Product Code. 10642243
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10642243 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10642243 Supplier Thermo Scientific Maybridge Supplier No. BTB12331EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 2725-81-7
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C9H5NO4
MDL Number MFCD00016973
Quantity 10 g
Synonym 6-nitrocoumarin, 6-nitro-2h-chromen-2-one, 6-nitro-2-benzopyrone, 2h-1-benzopyran-2-one, 6-nitro, acmc-20abd6, maybridge1_002226, amtgc084, 6-nitro-2h-chromen-2-one #, 2h-1-benzopyran-2-one,6-nitro, 6-hydroxy oxido amino-2h-chromen-2-one
InChI Key RMERXEXZXIVNBF-UHFFFAOYSA-N
SMILES C1=CC2=C(C=CC(=O)O2)C=C1[N+](=O)[O-]
IUPAC Name 6-nitrochromen-2-one
Molecular Weight (g/mol) 191.142
PubChem CID 75944
Formula Weight 191.14
Percent Purity 97%
Chemical Name or Material 6-Nitro-2H-chromen-2-one
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.